3-Acetyl-4'-(trifluoromethyl)biphenyl
Lieferant: Apollo Scientific
|
Achtung
|
PC11181-1G
APOSPC11181-1GEA
0
EUR
APOSPC11181-1G
3-Acetyl-4'-(trifluoromethyl)biphenyl
3-Acetyl-4'-(trifluoromethyl)biphenyl
Formel:
C₁₅H₁₁F₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801747 CAS-Nummer: 638214-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...