3'-(Trifluormethyl)biphenyl-3-carbonsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC11182-1G
PC11182-5G
APOSPC11182-1GEA
266
EUR
APOSPC11182-1G
APOSPC11182-5G
3'-(Trifluormethyl)biphenyl-3-carbonsäure 96%
3'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel:
C₁₄H₉F₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424603 CAS-Nummer: 168619-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...