4'-(Trifluormethyl)biphenyl-3-carbonsäure

Lieferant: Apollo Scientific
Achtung

PC11183-100MG PC11183-250MG
APOSPC11183-100MGEA 21.9 EUR
APOSPC11183-100MG APOSPC11183-250MG
4'-(Trifluormethyl)biphenyl-3-carbonsäure
4'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel: C₁₄H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03424604
CAS-Nummer: 199528-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR