1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC11195-1G
PC11195-250MG
PC11195-500MG
APOSPC11195-1GEA
348
EUR
APOSPC11195-1G
APOSPC11195-250MG
APOSPC11195-500MG
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperazine 95+%
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperazine
Formel:
C₁₁H₁₅FN₂O₂S Schmelzpunkt: 153…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037948 CAS-Nummer: 849938-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...