1,3-Dichlor-2-fluor-5-nitrobenzol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dichloro-4-fluoro-1-nitrobenzene
PC11210-25G
PC11210-100G
PC11210-500G
APOSPC11210-25GEA
21.9
EUR
APOSPC11210-25G
APOSPC11210-100G
APOSPC11210-500G
1,3-Dichlor-2-fluor-5-nitrobenzol 99%
1,3-Dichloro-2-fluoro-5-nitrobenzene
Formel:
C₆H₂Cl₂FNO₂ Molecular Weight: 209.99 g/mol Schmelzpunkt: 44 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129963 CAS-Nummer: 3107-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...