5-Fluorgramin 98%

Lieferant: Apollo Scientific

Synonyme: 1-(5-Fluoro-1H-indol-3-yl)-N,N-dimethylmethanamine

PC11211-1G
APOSPC11211-1GEA 87 EUR
APOSPC11211-1G
5-Fluorgramin 98%
5-Fluorgramin
Formel: C₁₁H₁₃FN₂
Molecular Weight: 192,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00022731
CAS-Nummer: 343-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR