2,3-Difluor-6-nitroanisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Difluoro-3-methoxy-4-nitrobenzene

PC11218-25G PC11218-5G
APOSPC11218-25GEA 40.6 EUR
APOSPC11218-25G APOSPC11218-5G
2,3-Difluor-6-nitroanisol
2,3-Difluor-6-nitroanisol
Formel: C₇H₅F₂NO₃
Molecular Weight: 189.1163 g/mol
Siedepunkt: 78…79 °C/20mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03840468
CAS-Nummer: 66684-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR