4-(3,5-Bis(trifluormethyl)phenyl)anilin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Amino-3',5'-bis(trifluoromethyl)biphenyl
PC11219-5G
APOSPC11219-5GEA
122
EUR
APOSPC11219-5G
4-(3,5-Bis(trifluormethyl)phenyl)anilin
4-(3,5-Bis(trifluormethyl)phenyl)anilin
Formel:
C₁₄H₉F₆N Lagertemperatur: Kühlschrank |
MDL:
MFCD05154989 CAS-Nummer: 444143-45-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...