Kalium(2-nitrophenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC11226-1G
PC11226-5G
PC11226-25G
APOSPC11226-1GEA
141
EUR
APOSPC11226-1G
APOSPC11226-5G
APOSPC11226-25G
Kalium(2-nitrophenyl)trifluoroborat 96%
Kalium(2-nitrophenyl)trifluoroborat
Formel:
C₆H₄BF₃KNO₂ Schmelzpunkt: 216…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115759 CAS-Nummer: 850623-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...