Kalium(3-chlorphenyl)trifluoroborat

Lieferant: Apollo Scientific

PC11229-1G PC11229-5G
APOSPC11229-1GEA 58.2 EUR
APOSPC11229-1G APOSPC11229-5G
Kalium(3-chlorphenyl)trifluoroborat
Kalium(3-chlorphenyl)trifluoroborat
Formel: C₆H₄BClF₃K
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115754
CAS-Nummer: 411206-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR