Kalium(4-ethoxyphenyl)trifluoroborat

Lieferant: Apollo Scientific

PC11230-1G PC11230-25G PC11230-5G
APOSPC11230-1GEA 121 EUR
APOSPC11230-1G APOSPC11230-25G APOSPC11230-5G
Kalium(4-ethoxyphenyl)trifluoroborat
Kalium(4-ethoxyphenyl)trifluoroborat
Formel: C₈H₉BF₃KO
Schmelzpunkt: 298…304 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115753
CAS-Nummer: 850623-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR