Kalium(3-chlor-4-fluorphenyl)trifluoroborat

Lieferant: Apollo Scientific

Synonyme: (3-Chlor-4-fluorphenyl)trifluorborsäure Kaliumsalz

PC11231-1G PC11231-5G PC11231-25G
APOSPC11231-1GEA 79.9 EUR
APOSPC11231-1G APOSPC11231-5G APOSPC11231-25G
Kalium(3-chlor-4-fluorphenyl)trifluoroborat
Kalium(3-chlor-4-fluorphenyl)trifluoroborat
Formel: C₆H₃BClF₄K
Molecular Weight: 236.44 g/mol
Schmelzpunkt: 234…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115752
CAS-Nummer: 850623-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR