Kalium(4-formylphenyl)trifluoroborat 98%

Lieferant: Apollo Scientific

PC11237-100G PC11237-25G
APOSPC11237-100GEA 0 EUR
APOSPC11237-100G APOSPC11237-25G
Kalium(4-formylphenyl)trifluoroborat 98%
Kalium(4-formylphenyl)trifluoroborat
Formel: C₇H₅BF₃KO
Schmelzpunkt: 282…288 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093980
CAS-Nummer: 374564-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR