Kalium(4-(benzyloxy)phenyl)trifluoroborat
Lieferant: Apollo Scientific
Synonyme:
Potassium (4-(benzyloxy)phenyl)trifluoroborate
PC11238-5G
APOSPC11238-5GEA
115
EUR
APOSPC11238-5G
Kalium(4-(benzyloxy)phenyl)trifluoroborat
Kalium(4-(benzyloxy)phenyl)trifluoroborat
Formel:
C₁₃H₁₁BF₃KO Molecular Weight: 290.13 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04115746 CAS-Nummer: 850623-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...