Kalium(3-cyanphenyl)trifluoroborat
Lieferant: Apollo Scientific
Synonyme:
(3-Cyanophenyl)trifluoroboric acid potassium salt
PC11239-25G
APOSPC11239-25GEA
140
EUR
APOSPC11239-25G
Kalium(3-cyanphenyl)trifluoroborat
Kalium(3-cyanphenyl)trifluoroborat
Formel:
C₇H₄BF₃KN Molecular Weight: 209.02 g/mol Schmelzpunkt: 196…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115744 CAS-Nummer: 850623-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...