Kalium(4-(methylsulfonyl)phenyl)trifluoroborat

Lieferant: Apollo Scientific

PC11244-1G PC11244-5G
APOSPC11244-1GEA 86.1 EUR
APOSPC11244-1G APOSPC11244-5G
Kalium(4-(methylsulfonyl)phenyl)trifluoroborat
Potassium-4-(methylsulphonyl)phenyltrifluoroborate
Formel: C₇H₇BF₃KO₂S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115738
CAS-Nummer: 850623-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR