Kalium(4-cyanphenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Cyanophenyl)trifluoroboric acid potassium salt
PC11246-25G
APOSPC11246-25GEA
183
EUR
APOSPC11246-25G
Kalium(4-cyanphenyl)trifluoroborat 96%
Kalium(4-cyanphenyl)trifluoroborat
Formel:
C₇H₄BF₃KN Molecular Weight: 209.02 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115736 CAS-Nummer: 850623-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...