Kalium(4-cyanphenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC11246-1G
PC11246-5G
PC11246-25G
APOSPC11246-1GEA
0
EUR
APOSPC11246-1G
APOSPC11246-5G
APOSPC11246-25G
Kalium(4-cyanphenyl)trifluoroborat 96%
Kalium(4-cyanphenyl)trifluoroborat
Formel:
C₇H₄BF₃KN Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115736 CAS-Nummer: 850623-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...