2-Chloro-N-(2,3,4,5,6-pentafluorophenyl)acetamide
Lieferant: Apollo Scientific
Synonyme:
N-(Chloroacetyl)pentafluoroaniline
PC11249-1G
APOSPC11249-1GEA
93.1
EUR
APOSPC11249-1G
2-Chloro-N-(2,3,4,5,6-pentafluorophenyl)acetamide
2-Chloro-N-(2,3,4,5,6-pentafluorophenyl)acetamide
Formel:
C₈H₃ClF₅NO Schmelzpunkt: 105…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077475 CAS-Nummer: 70426-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...