2-Chloro-N-(2,4,5-trifluorophenyl)acetamide

Lieferant: Apollo Scientific

Synonyme: N-(Chloroacetyl)-2,4,5-trifluoroaniline

PC11250-1G
APOSPC11250-1GEA 180 EUR
APOSPC11250-1G
2-Chloro-N-(2,4,5-trifluorophenyl)acetamide
2-Chloro-N-(2,4,5-trifluorophenyl)acetamide
Formel: C₈H₅ClF₃NO
Molecular Weight: 223,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077477
CAS-Nummer: 885267-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR