2-Chloro-N-(5-fluoro-2-methylphenyl)acetamide

Lieferant: Apollo Scientific

Synonyme: N-(Chloroacetyl)-5-fluoro-2-methylaniline

PC11252-1G
APOSPC11252-1GEA 93.1 EUR
APOSPC11252-1G
2-Chloro-N-(5-fluoro-2-methylphenyl)acetamide
2-Chloro-N-(5-fluoro-2-methylphenyl)acetamide
Formel: C₉H₉ClFNO
Molecular Weight: 201,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788386
CAS-Nummer: 85817-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR