3-Brom-4-hydroxy-2-(trifluormethyl)chinolin

Lieferant: Apollo Scientific
Achtung

PC11262-1G
APOSPC11262-1GEA 29 EUR
APOSPC11262-1G
3-Brom-4-hydroxy-2-(trifluormethyl)chinolin
3-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline
Formel: C₁₀H₅BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD03407378
CAS-Nummer: 59108-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR