2-Chlor-4-(trifluormethyl)chinolin
Lieferant: Apollo Scientific
PC11266-1G
APOSPC11266-1GEA
277
EUR
APOSPC11266-1G
2-Chlor-4-(trifluormethyl)chinolin
2-Chloro-4-(trifluoromethyl)quinoline
Formel:
C₁₀H₅ClF₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407381 CAS-Nummer: 2806-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...