N,N-Dimethyl-2-(trifluoromethoxy)benzylamine

Lieferant: Apollo Scientific
Achtung

PC11281-25G PC11281-5G
APOSPC11281-25GEA 0 EUR
APOSPC11281-25G APOSPC11281-5G
N,N-Dimethyl-2-(trifluoromethoxy)benzylamine
N,N-Dimethyl-2-(trifluoromethoxy)benzylamine
Formel: C₁₀H₁₂F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07368741
CAS-Nummer: 886762-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR