3,6-Difluoro-2-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,6-Difluoro-o-anisic acid, 2-Carboxy-3,6-difluoroanisole

PC1138-1G PC1138-5G
APOSPC1138-1GEA 43.5 EUR
APOSPC1138-1G APOSPC1138-5G
3,6-Difluoro-2-methoxybenzoic acid
3,6-Difluoro-2-methoxybenzoic acid
Formel: C₈H₆F₂O₃
Molecular Weight: 188.128 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08275290
CAS-Nummer: 887267-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR