3,6-Difluoro-2-methoxybenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,6-Difluoro-o-anisic acid, 2-Carboxy-3,6-difluoroanisole
PC1138-1G
PC1138-5G
APOSPC1138-1GEA
43.5
EUR
APOSPC1138-1G
APOSPC1138-5G
3,6-Difluoro-2-methoxybenzoic acid
3,6-Difluoro-2-methoxybenzoic acid
Formel:
C₈H₆F₂O₃ Molecular Weight: 188.128 g/mol Schmelzpunkt: 85…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275290 CAS-Nummer: 887267-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...