(S)-N-Methyl-1-[3,5-bis(trifluoromethyl)phenyl]ethylamine

Lieferant: Apollo Scientific
Achtung

PC1166-1G
APOSPC1166-1GEA 325 EUR
APOSPC1166-1G
(S)-N-Methyl-1-[3,5-bis(trifluoromethyl)phenyl]ethylamine
(S)-N-Methyl-1-[3,5-bis(trifluoromethyl)phenyl]ethylamine
Formel: C₁₁H₁₁F₆N
Molecular Weight: 271,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779108
CAS-Nummer: 511256-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR