4-(2-Cyanoethyl)-5-(trifluoromethyl)isoxazole

Lieferant: Apollo Scientific

PC1183-500MG
APOSPC1183-500MGEA 274 EUR
APOSPC1183-500MG
4-(2-Cyanoethyl)-5-(trifluoromethyl)isoxazole
4-(2-Cyanoethyl)-5-(trifluoromethyl)isoxazole
Formel: C₇H₅F₃N₂O
Molecular Weight: 190,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07787362
CAS-Nummer: 220370-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR