Benzotrifluorid 99%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α,α,α-Trifluortoluol, Lösungsmittel BTF
PC1190-1KG
APOSPC1190-1KGEA
346
EUR
APOSPC1190-1KG
Benzotrifluorid 99%
Benzotrifluorid
An alternative solvent to dichloromethane with higher bp and lower toxicity.
Formel:
C₇H₅F₃ Molecular Weight: 146,11 g/mol Siedepunkt: 101…102 °C Schmelzpunkt: –29 °C Dichte: 1,199 g/cm³ (25 °C) Flammpunkt: 12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000372 CAS-Nummer: 98-08-8 EINECS: 202-635-0 UN: 2338 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...