5-Aminomethyl-3-isopropylisoxazoletrifluoroacetate
Lieferant: Apollo Scientific
PC1195-250MG
APOSPC1195-250MGEA
288
EUR
APOSPC1195-250MG
5-Aminomethyl-3-isopropylisoxazoletrifluoroacetate
5-Aminomethyl-3-isopropylisoxazoletrifluoroacetate
Formel:
C₉H₁₃F₃N₂O₃ Molecular Weight: 254.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953735 CAS-Nummer: 1159599-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...