3,4-Difluor-5-methoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (3,4-Difluoro-5-methoxyphenyl)boronic acid

PC1202-1G PC1202-25G PC1202-5G
APOSPC1202-1GEA 119 EUR
APOSPC1202-1G APOSPC1202-25G APOSPC1202-5G
3,4-Difluor-5-methoxyphenylboronsäure
3,4-Difluor-5-methoxyphenylboronsäure
Formel: C₇H₇BF₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD09907960
CAS-Nummer: 925910-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR