4-Brom-2,6-difluorphenol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-3,5-difluoro-4-hydroxybenzene
PC1209-1G
PC1209-5G
PC1209-10G
APOSPC1209-1GEA
21.9
EUR
APOSPC1209-1G
APOSPC1209-5G
APOSPC1209-10G
4-Brom-2,6-difluorphenol 98%
4-Brom-2,6-difluorphenol
Formel:
C₆H₃BrF₂O Schmelzpunkt: 50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236213 CAS-Nummer: 104197-13-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...