Perfluoro(3,7-dimethyloctanoic acid)

Lieferant: Apollo Scientific
Gefahr

Synonyme: Nonadecafluoro(3,7-dimethyloctanoic acid), 3,7-Bis(trifluoromethyl)-2,2,3,4,4,5,5,6,6,7,8,8,8-tridecafluorooctanoic acid

PC1214-25G PC1214-5G PC1214-1G
APOSPC1214-25GEA 760 EUR
APOSPC1214-25G APOSPC1214-5G APOSPC1214-1G
Perfluoro(3,7-dimethyloctanoic acid)
Perfluoro(3,7-dimethyloctanoic acid)

Liquid at room temperature

Formel: C₁₀HF₁₉O₂
Molecular Weight: 514.083 g/mol
Siedepunkt: 95 °C (20 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01320738
CAS-Nummer: 172155-07-6
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR