2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane 99%

Lieferant: Apollo Scientific
Achtung

PC1224-100G PC1224-250G PC1224-500G
APOSPC1224-100GEA 109 EUR
APOSPC1224-100G APOSPC1224-250G APOSPC1224-500G
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane 99%
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane

  • Polymer grade

Formel: C₁₅H₁₂F₆N₂O₂
Schmelzpunkt: 243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039147
CAS-Nummer: 83558-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR