2,2-Bis(4-methylphenyl)hexafluoropropane 98%
Lieferant: Apollo Scientific
Synonyme:
4,4'-(Hexafluoroisopropylidene)ditoluene, 4,4'-(Perfluoropropane-2,2-diyl)bis(methylbenzene)
PC1230-25G
APOSPC1230-25GEA
59.6
EUR
APOSPC1230-25G
2,2-Bis(4-methylphenyl)hexafluoropropane 98%
2,2-Bis(4-methylphenyl)hexafluoropropane
Formel:
C₁₇H₁₄F₆ Molecular Weight: 332,29 g/mol Schmelzpunkt: 82…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042597 CAS-Nummer: 1095-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...