3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine, 5,6,7,8-Tetrahydro-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyrazine

PC1232-1G PC1232-250MG PC1232-5G
APOSPC1232-1GEA 36.4 EUR
APOSPC1232-1G APOSPC1232-250MG APOSPC1232-5G
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine
Formel: C₆H₇F₃N₄
Molecular Weight: 192.145 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08460920
CAS-Nummer: 486460-21-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR