2-Fluor-5-hydroxypyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1240-25G
PC1240-5G
PC1240-100G
APOSPC1240-25GEA
91.5
EUR
APOSPC1240-25G
APOSPC1240-5G
APOSPC1240-100G
2-Fluor-5-hydroxypyridin 98%
2-Fluor-5-hydroxypyridin
Formel:
C₅H₄FNO Schmelzpunkt: 145…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075040 CAS-Nummer: 55758-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...