(S)-(+)-1-Boc-3-fluoropyrrolidine 97%

Lieferant: Apollo Scientific

Synonyme: (3S)-3-Fluoropyrrolidine, N-BOC protected, (S)-N-boc-3-fluoropyrrolidine

PC1252-250MG
APOSPC1252-250MGEA 75.4 EUR
APOSPC1252-250MG
(S)-(+)-1-Boc-3-fluoropyrrolidine 97%
(S)-(+)-1-Boc-3-fluoropyrrolidine
Formel: C₉H₁₆FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04114064
CAS-Nummer: 479253-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR