N, O-Bis (Trifluoracetyl)Hydroxylamin 95%

Lieferant: Apollo Scientific

PC1252G-5G
APOSPC1252G-5GEA 328 EUR
APOSPC1252G-5G
N, O-Bis (Trifluoracetyl)Hydroxylamin 95%
N, O-Bis (Trifluoracetyl)Hydroxylamin
Formel: C₄HF₆NO₃
Molecular Weight: 225,05 g/mol
Schmelzpunkt: 59…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013565
CAS-Nummer: 684-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR