n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal 95%

Lieferant: Apollo Scientific

Synonyme: Acetaldehyde butyl 2,2,2-trifluoroethyl acetal

PC1267-100G PC1267-25G
APOSPC1267-100GEA 911 EUR
APOSPC1267-100G APOSPC1267-25G
n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal 95%
n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal
Formel: C₈H₁₅F₃O₂
Molecular Weight: 200.2 g/mol
Siedepunkt: 143.2…143.3 °C/751mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03094109
CAS-Nummer: 2925-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR