Perfluoro(1-ethyl-3-propoxycyclohexane) 97%

Lieferant: Apollo Scientific

PC1294-1G
APOSPC1294-1GEA 170 EUR
APOSPC1294-1G
Perfluoro(1-ethyl-3-propoxycyclohexane) 97%
Perfluoro(1-ethyl-3-propoxycyclohexane)
Formel: C₁₁F₂₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03094121
CAS-Nummer: 400626-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR