2-(4-Fluorphenoxy)nicotinsäure 97%

Lieferant: Apollo Scientific
Achtung

PC1298-10G
APOSPC1298-10GEA 147 EUR
APOSPC1298-10G
2-(4-Fluorphenoxy)nicotinsäure 97%
2-(4-Fluorphenoxy)nicotinsäure
Formel: C₁₂H₈FNO₃
Molecular Weight: 233,20 g/mol
Schmelzpunkt: 179…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00833409
CAS-Nummer: 54629-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR