2,4-Difluorophenylurea

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Carbamoylamino)-2,4-difluorobenzene, 2,4-Difluoro-1-ureidobenzene

PC1316-1G
APOSPC1316-1GEA 219 EUR
APOSPC1316-1G
2,4-Difluorophenylurea
2,4-Difluorophenylurea
Formel: C₇H₆F₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00025401
CAS-Nummer: 145602-64-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR