3-Chlor-1-fluor-4-nitrobenzol
Lieferant: Apollo Scientific
|
Achtung
|
PC1336-1G
PC1336-25G
PC1336-5G
APOSPC1336-1GEA
0
EUR
APOSPC1336-1G
APOSPC1336-25G
APOSPC1336-5G
3-Chlor-1-fluor-4-nitrobenzol
3-Chlor-1-fluor-4-nitrobenzol
Formel:
C₆H₃ClFNO₂ Schmelzpunkt: 33…37 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412200 CAS-Nummer: 2106-50-5 EINECS: 218-286-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...