4-Brom-3-(trifluormethyl)benzonitril 97%

Lieferant: Apollo Scientific
Achtung

PC1337-25G PC1337-5G
APOSPC1337-25GEA 37.7 EUR
APOSPC1337-25G APOSPC1337-5G
4-Brom-3-(trifluormethyl)benzonitril 97%
4-Brom-3-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
Molecular Weight: 250.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095347
CAS-Nummer: 1735-53-1
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR