4-Fluor-1,3-benzoldiamin 98%
Lieferant: Apollo Scientific
Synonyme:
1,3-Diamino-4-fluorobenzene, 4-Fluorobenzene-1,3-diamine
PC1338-5G
APOSPC1338-5GEA
116
EUR
APOSPC1338-5G
4-Fluor-1,3-benzoldiamin 98%
4-Fluor-1,3-benzoldiamin
Formel:
C₆H₇FN₂ Molecular Weight: 126,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077494 CAS-Nummer: 6264-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...