2-Chlor-3-fluortoluol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chlor-2-fluor-6-methylbenzol

PC1360-5G
APOSPC1360-5GEA 127 EUR
APOSPC1360-5G
2-Chlor-3-fluortoluol 98%
2-Chlor-3-fluortoluol
Formel: C₇H₆ClF
Molecular Weight: 144.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445246
CAS-Nummer: 116850-28-3
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR