2-Brom-4,5-difluorbenzonitril

Lieferant: Apollo Scientific
Achtung

PC1369-25G PC1369-5G
APOSPC1369-25GEA 74.1 EUR
APOSPC1369-25G APOSPC1369-5G
2-Brom-4,5-difluorbenzonitril
2-Brom-4,5-difluorbenzonitril
Formel: C₇H₂BrF₂N
Schmelzpunkt: 63…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09864661
CAS-Nummer: 64695-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR