2-Brom-3-chlor-5-(trifluormethyl)pyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1374E-1G
APOSPC1374E-1GEA
21.9
EUR
APOSPC1374E-1G
2-Brom-3-chlor-5-(trifluormethyl)pyridin 97%
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Formel:
C₆H₂BrClF₃N Siedepunkt: 80 °C/32mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153072 CAS-Nummer: 75806-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...