5-Bromo-4-chloro-4,5,5-trifluoropentanoicacid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1374KE-250MG
APOSPC1374KE-250MGEA
69.6
EUR
APOSPC1374KE-250MG
5-Bromo-4-chloro-4,5,5-trifluoropentanoicacid 97%
5-Bromo-4-chloro-4,5,5-trifluoropentanoicacid
Formel:
C₅H₅BrClF₃O₂ Molecular Weight: 269.45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153680 CAS-Nummer: 232602-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...